Learn More
Sigma Aldrich Fine Chemicals Biosciences IgM Lambda from murine mye1MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M51701MG
Encompass
IgM antibodies are present as pentamers in the serum and are produced in response to antigens IgM » from murine myeloma is produced by MOPC 104E tumor line introduced subcutaneously in BALB/c mice. It specifically recognizes mouse IgM ». Identity and purity of the immunoglobulin is established by immunoelectrophoresis.
Catalog No. 50-290-4502