Learn More
Cayman Chemical IgMhumn Monclonal ntIbo 100ug
SDP
Supplier:  Cayman Chemical 32113100 UG
Encompass_Preferred
Immunogen: Human IgM • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human IgM; (-) Human IgG, IgA, IgD, or IgE • Applications: ELISA, ICC, IHC
Catalog No. 50-294-6530