Learn More
Selleck Chemical LLC iGOT1-01 S4455-100mg
SDP
Supplier:  Selleck Chemical LLC S4455100MG
Encompass_Preferred
iGOT1-01 a potent aspartate aminotransferase 1 (glutamate oxaloacetate transaminase 1 GOT1) inhibitor possesses anti-cancer activity
Catalog No. 50-311-0492