Learn More
Ihc World LLC IHC TEK DAPI SOLUTION 50ML
SDP
Supplier:  Ihc World LLC IW1404
Encompass_Preferred
This DAPI Counterstain Solution is designed for nuclear counterstain after completion of immunofluorescent staining. Its blue fluorescence is in excellent contrast to green and red fluorescent probes such as FITC and Texas Red. This DAPI Solution is suitable for both immunofluorescent counterstain of tissue sections and cultured cells grown on slides as well as whole mount staining.
Catalog No. NC0056218