Learn More
Medchemexpress LLC IHMT-MST1-58 | 2414484-25-4 | C21H22N6O3S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15125710MM/1ML
Encompass_Preferred
IHMT-MST1-58 is a potent, selective, mammalian, and orally active STE20-like protein 1 kinase (MST1) inhibitor with an IC50 value of 23 nM. It is utilized in research for type 1/2 diabetes.
  • Potent MST1 inhibitor with an IC50 of 23 nM.
  • Weak activity against MST2 and no activity against NEK3.
  • Strong binding affinity to MST1.
  • Inhibits phosphorylation of MST1 and downstream MOB1 autophosphorylation.
  • Provides protective effect for β cells against inflammatory cytokine damage.
  • Oral administration decreased fasting blood glucose and HbA1c levels in STZ-induced T1D/T2D mouse models.
  • Acceptable pharmacokinetic properties across species.
  • Purity: 99.05%.
Catalog No. 50-003-75827