Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC iHSP110-33 | 99.59% | 574.67 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY161765

Encompass_Preferred

iHSP110-33 is an inhibitor for heat shock protein 110 (HSP110). It exhibits antitumor efficacy against large B-cell lymphoma and classical Hodgkin lymphoma. iHSP110-33 shows a synergistic effect with Selinexor (HY-17536), inhibits the STAT6 phosphorylation, and enhances its antitumor activity.

  • Solution: 10 mM * 1 mL in DMSO
  • Solid: 5 mg
  • Solid: 10 mg
  • Solid: 25 mg
  • Solid: 50 mg
  • Solid: 100 mg
  • Solid: 200 mg (Get quote)
  • Solid: 500 mg (Get quote)

Catalog No. 50-003-95222


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.