Learn More
Cayman Chemical IKBaPhospho-Ser3236 Monc 1ea
SDP
Supplier:  Cayman Chemical 139231 EA
Encompass_Preferred
Immunogen: A synthetic peptide containing phosphorylated serines at amino acid residues 32 and 36 of human IκBα • Host: Mouse • Species Reactivity: (+) Human, mouse, rat, porcine, bovine, dog • Cross Reactivity: (+) IκBα (phospho-Ser32/36) • Applications: IP, WB
Catalog No. 50-295-5294