Learn More
Medchemexpress LLC HY-124952 5mg Medchemexpress, iKIX1 CAS:656222-54-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1249525MG
Encompass_Preferred
Medchemexpress, HY-124952 5mg iKIX1 CAS:656222-54-7 iKIX1 is an antifungal agent and resensitizes drug-resistant C. glabrata to azole antifungals in vitro. iKIX1 inhibits the interaction between the KIX domain of the mediator subunit CgGal11A and the activation domain of CgPdr1, the IC50 and Ki values are 190.2 μM and 18 μM, respectively. iKIX1 is used for the study of multidrug resistance and C. glabrata infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1060