Learn More
TARGETMOL CHEMICALS INC IKK 16 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T617610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IKK 16 (IKK Inhibitor VII) is a selective I(kappa)B kinase (IKK) inhibitor for IKK-2 IKK complex and IKK-1 with IC50 of 40 nM 70 nM and 200 nM respectively. purity: 99%
Catalog No. 50-257-9503