Learn More
Selleck Chemical LLC IKK-16 (IKK Inhibitor VII) 10mM/1mL 873225-46-8
SDP
Supplier:  Selleck Chemical LLC S288210MM/1ML
Encompass_Preferred
IKK-16 (IKK Inhibitor VII) is a selective I?B kinase (IKK) inhibitor for IKK-2, IKK complex and IKK-1 with IC50 of 40 nM, 70 nM and 200 nM in cell-free assays, respectively. IKK-16 also inhibits LRRK2 Ser935 phosphorylation in cells and LRRK2 kinase activity in vitro with IC50 of 50 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3129