Learn More
ABclonal Technology IKK Rabbit mAb
SDP
Supplier:  ABclonal Technology A1960620UL
Encompass_Preferred
The protein encoded by this gene phosphorylates the inhibitor in the inhibitor NF-kappa-B complex causing dissociation of the inhibitor and activation of NF-kappa-B The encoded protein itself is found in a complex of proteins Several transcript variants some protein-coding and some not have been found for this gene
Catalog No. 50-000-08303