Learn More
Cayman Chemical IKK2 InhIbItr VI 5mg
SDP
Supplier:  Cayman Chemical 172765 MG
Encompass_Preferred
A potent, cell-permeable, reversible inhibitor of IKK2 (IC50 = 13 nM); used to evaluate the role of the canonical, IκB-dependent NF-κB signaling pathway in cellular responses
Catalog No. 50-297-9643