Learn More
Medchemexpress LLC IL-1 alpha/IL-1F1 P 10ug | 10ug
SDP
Supplier:  Medchemexpress LLC HYP7924310UG
Encompass_Preferred
The IL-1 /IL-1F1 protein is found intracellularly in most non-hematopoietic cells and plays a crucial role in mediating inflammation and linking innate and adaptive immunity IL1RAP binds to IL1R1 to form a high-affinity receptor complex that activates cascades and pathways IL-1 alpha/IL-1F1 Protein Pig is the recombinant Porcine-derived IL-1 alpha/IL-1F1 protein expressed by E coli with tag free
Catalog No. 50-001-94676