Learn More
Medchemexpress LLC IL-1 beta Cavia por 50ug
SDP
Supplier:  Medchemexpress LLC HYP7257150UG
Encompass_Preferred
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Cavia porcellus (His) is produced in E coli with six C-Terminal His-tags It consists of 152 amino acids (T115-S266)
Catalog No. 50-001-82505