Learn More
Medchemexpress LLC IL-1 beta Human C- 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP78459A50UG
Encompass_Preferred
IL-1 beta protein is a potent proinflammatory cytokine that plays multiple roles in immune responses It induces inflammatory events including prostaglandin synthesis neutrophil activation and cytokine production IL-1 beta Protein Human (C-His) is the recombinant human-derived IL-1 beta protein expressed by E coli with C-6 His labeled tag
Catalog No. 50-001-94445