Learn More
Medchemexpress LLC IL-1 beta Human CH 2ug
SDP
Supplier:  Medchemexpress LLC HYP72052UG
Encompass_Preferred
Recombinant human interleukin-1 beta (IL-1β) expressed in Chinese hamster ovary (CHO) cells and supplied lyophilized for research use. IL-1β is a potent proinflammatory cytokine commonly used in vitro to study immune signaling, inflammation, and cytokine responses. The product is high purity (>95%) with a predicted molecular mass of ~17 kDa and is formulated after dialysis to preserve activity.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Predicted molecular mass of approximately 17 kDa.
  • Lyophilized formulation for convenient storage and transport.
  • Formulated after extensive dialysis in physiological buffer to preserve activity.
  • Stable when stored lyophilized at -20°C; aliquot and store at -20°C or -80°C for extended stability.
  • Suitable for in vitro bioassays and signaling studies of inflammatory pathways.
Catalog No. 50-001-95479