Learn More
Medchemexpress LLC IL-1 beta Human Hi 500ug | 500ug
SDP
Supplier:  Medchemexpress LLC HYP78459500UG
Encompass_Preferred
IL-1 beta is a potent proinflammatory cytokine expressed by monocytes macrophages and dendritic cells IL-1 beta plays a key role in inflammation and immunologic reactions[1][2] IL-1 beta Protein Human (His-Avi) is produced in E coli with C-Terminal 6 His-tag and a C-Terminal Avi-tag It consists of 153 amino acids (A117-S269)
Catalog No. 50-001-94451