Learn More
Medchemexpress LLC IL-10R beta Human 2ug
SDP
Supplier:  Medchemexpress LLC HYP725992UG
Encompass_Preferred
IL-10R beta protein is an IL10 cytokine surface receptor involved in IL10-mediated inflammation and immune regulation as well as viral defense responses IL-10R beta Protein Human (HEK293 His) is expressed by HEK 293 cells and has a transmembrane region (W221-F242) with a 6 His tag at the C-terminus[3]
Catalog No. 50-001-82595