Learn More
Medchemexpress LLC IL-12 beta Rat HEK 50ug
SDP
Supplier:  Medchemexpress LLC HYP7316450UG
Encompass_Preferred
IL-12 beta Protein also known as natural killer cell stimulatory factor 2 is a common subunit (p40) of IL-12 and IL-23 IL-12 is a inflammatory factor expressed by activated macrophages and involves in Th1-type immune response against cancer[1] IL-12 beta Protein located outside the cell membrane involves in singalling mediated by Jak-STAT[3] IL-12 beta Protein consists of 335 amino acids (M1-S335) with a lg-like domain (43-90 a a) IL-12 beta Protein Rat (M1-S335) is produced in HEK293 cells with a C-Terminal His-tag
Catalog No. 50-001-82401