Learn More
ACROBiosystems ACROBiosystems Human IL-13 Protein, His Tag
Supplier:  ACROBiosystems IL3H52H450UG
Encompass_Preferred
Recombinant Protein;50UG;Human IL-13, His Tag (IL3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Asn 132 (Accession # AAK53823.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.2 kDa. The protein migrates as 15 kDa and 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse IL-13RA1, Fc Tag (Cat. No. IL1-M5258) at 2 μg/mL (100 μL/well) can bind Human IL-13, His Tag (Cat. No. IL3-H52H4) with a linear range of 5-39 ng/mL (QC tested).;Immobilized Human IL-13, His Tag (Cat. No. IL3-H52H4) at 5 μg/mL (100 μL/well) can bind Monoclonal Anti-Human IL-13 Antibody, Human IgG4 with a linear range of 0.3-5 ng/mL (Routinely tested).;Human IL-13, His Tag (Cat. No. IL3-H52H4) stimulates proliferation of TF-1 cells. The EC50 for this effect is 0.05-0.08 μg/mL (Routinely tested).
Catalog No. 50-195-6789