Learn More
Enzo Life Sciences IL-13 (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1830010
Encompass_Preferred
Alternative name: Interleukin-13, NC30. Purity: ≥98% (SDS & HPLC). Formulation: Lyophilized from 20mM sodium phosphate, pH 7.0. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-3970