Learn More
Eurofins Discoverx Recombinant Human IL-17 (IL-17A), 25 µg. (92-1229)
Supplier:  Eurofins Discoverx 921229
Encompass
Interleukin 17A (IL-17, IL-17A) is a pro-inflammatory cytokine protein produced by activated T cells and the founding member of the IL-17 family. Potent mediator in delayed-type inflammatory reactions that increases chemokine production and recruits monocytes and neutrophils to sites of inflammation. Agonist of the interleukin receptor IL-17RA. High levels are associated with several chronic inflammatory diseases, such as rheumatoid arthritis, psoriasis, and multiple sclerosis. Provided as 25 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9752