Learn More
Medchemexpress LLC Il-17 modulator 1 disodium | 2446803-91-2 | 98.4% | C28H35N6Na2O6P | 1 ML
SDP
Supplier:  Medchemexpress LLC HY141535A10MM/1ML
Encompass_Preferred
IL-17 modulator 1 disodium is an orally active, highly efficacious small molecule IL-17 modulator. Extracted from patent WO 2020127685, this salt form offers enhanced water solubility and stability compared to its free form. It is suitable for research aimed at preventing or ameliorating diseases such as psoriasis, ankylosing spondylitis, and psoriatic arthritis. This product is for research use only.
  • Orally active and highly efficacious
  • Small molecule IL-17 modulator
  • Enhanced water solubility and stability in disodium salt form
  • Useful for research into psoriasis, ankylosing spondylitis, and psoriatic arthritis
  • White to off-white solid appearance
Catalog No. 50-003-53763