Learn More
Enzo Life Sciences IL-17A (human), (recombinant) (25µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1880025
Encompass_Preferred
Alternative name: Interleukin-17, CTLA8, Cytotoxic T-lymphocyte-associated serine esterase 8. Purity: ≥98% (SDS & HPLC). Formulation: Lyophilized with no additives. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-3975