Learn More
Medchemexpress LLC HY-139206 1mg , IL-17A inhibitor 1 CAS:2452464-73-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1392061MG
Encompass_Preferred
HY-139206 1mg IL-17A inhibitor 1 CAS: 2452464-73-0 IL-17A inhibitor 1 (example 24) is a IL-17A inhibitor, with IC50 values of <9.45 nM and 9.3 nM in alphalisa assay and HT-29 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1818