Learn More
Medchemexpress LLC IL-17A inhibitor 1 | 2452464-73-0 | MFCD34474218 | >98.0% | C24H27F5N8O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13920610MG
Encompass_Preferred
LY3509754 is an orally active small-molecule inhibitor of interleukin-17A (IL-17A) developed by Eli Lilly. It exhibits low-nanomolar potency in biochemical and cell-based assays, has been evaluated in early-phase clinical studies, and is supplied for research use with documented purity and supporting analytical data.
  • Potent IL-17A inhibition with low-nanomolar IC50 values.
  • Demonstrated cellular activity in HT-29 and NHDF assays.
  • Supplied for research use with documented purity greater than 98.0%.
  • Orally active small-molecule chemical class appropriate for pharmacology studies.
  • Supported by preclinical and Phase 1 clinical data for translational research.
Catalog No. 50-002-61207