Learn More
Medchemexpress LLC IL-18 Rat 10ug
SDP
Supplier:  Medchemexpress LLC HYP721010UG
Encompass_Preferred
Recombinant rat interleukin-18 (IL-18), produced in E. coli and supplied lyophilized for research use. It activates interferon-γ production and is suitable for cell-based bioactivity assays and functional studies of IL-18 signaling.
  • Stimulates interferon-γ production in target cells.
  • Produced in E. coli expression system for consistent quality.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Approximately 18.4 kDa molecular weight as assessed by SDS-PAGE.
  • Biological activity greater than 500 units/mg in KG-1 cell assay.
  • Stable at -20°C for long-term storage; reconstitute and aliquot for use.
Catalog No. 50-001-95439