Learn More
Enzo Life Sciences IL-1 beta (human), (recombinant) (cell culture grade) (10µg). CAS: 97599-21-8
SDP
Supplier:  Enzo Life Sciences ALX520001C010
Encompass_Preferred
Potent immunomodulator. Mediates a wide range of immune and inflammatory responses including the activation of B and T cells. Potent immunomodulator. Mediates a wide range of immune and inflammatory responses including the activation of B and T cells. Alternative name: IL-1F2, Interleukin-1 beta. Purity: ≥97% (SDS-PAGE). Formulation: Lyophilized. Salt-free. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-200-9347