Learn More
Medchemexpress LLC IL-2-IN-1 | 245747-10-8 | 99.9% | 418.29 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1500515MG
Encompass_Preferred
IL-2-IN-1 is a potent IL-2 inhibitor with an IC50 value of 1978 nM. It exhibits antiproliferative activities. This product is for research use only.
  • Targets IL-2
  • Has an IC50 of 1978 nM
  • Inhibits antiproliferative activities in human PBMC with an IC50 of 340 nM
  • Inhibits antiproliferative activities in rat PBMC with an IC50 of 1266 nM
Catalog No. 50-003-60199