Learn More
AdipoGen IL-2 Superkine Fc
SDP
Supplier:  AdipoGen AG40B01113010
Encompass_Preferred
Protein. The extracellular domain of human IL-2 aa 21-153 mutant H9 containing the mutations L80F / R81D / L85V / I 86V / I92F is fused at the C-terminus to the Fc portion of human IgG2. Source HEK 293 cells. Endotoxin content <0.01EU/µg protein LAL test Lonza. Lyophilized. Contains PBS. Binds to human and mouse IL-2R. Purity >95% SDS-PAGE. Interleukin-2 IL-2 is a 133 amino acid glycoprotein with one intramolecular disulfide bond and variable glycosylation. It is secreted by activated T cells and induces proliferation and maturation of activated T cells, natural killer cells and lymphokine activated killer cells. IL-2 also stimulates proliferation of antibody-producing B cells, activates neutrophils and induces mononuclear cells to secrete IFN-gamma and TNF-alpha and -beta.
Catalog No. 50-256-2917