Learn More
Medchemexpress LLC IL-21 Mouse 129a.a 2ug

Supplier: Medchemexpress LLC HYP70782UG
Recombinant mouse interleukin-21 (IL-21) protein, 129 amino acids, supplied as a tag-free, lyophilized product expressed in E. coli. It is provided at high purity for use in immunology and cell biology research, with recommended storage and reconstitution guidance for experimental stability.
- Tag free, recombinant protein expressed in E. coli.
- Mature sequence of 129 amino acids (H18-S146).
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Lyophilized after dialysis in PBS for enhanced stability.
- Approximate molecular weight ~15.1 kDa.
- Includes storage and reconstitution recommendations for short and long term use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.