Learn More
Medchemexpress LLC IL-21R Mouse HEK29 100ug
SDP
Supplier:  Medchemexpress LLC HYP73889100UG
Encompass_Preferred
The IL-21R protein is specifically designed for IL-21 and forms a heterodimeric complex with the common subunit which is critical for signal transduction This receptor complex promotes interaction with Janus kinase 1 (JAK1) initiating downstream signaling events in response to interleukin-21 IL-21R Protein Mouse (HEK293 Fc) is the recombinant mouse-derived IL-21R protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-79310