Learn More
Medchemexpress LLC Interleukin 23 | >90.0% | 20UG

Supplier: Medchemexpress LLC HYP70074520UG
Recombinant human interleukin-23 (heterodimer of p19 and p40), biotinylated and produced in HEK293 cells. Supplied as a 0.2 μm filtered PBS solution, this protein is designed for research into IL-23 receptor binding, JAK/STAT signaling, and Th17-related immune responses. Handle and store according to supplier recommendations to maintain activity.
- Human IL-23 heterodimer (p19 + p40).
- Biotinylated for affinity-based detection and capture.
- Expressed in HEK293 mammalian cells for native glycosylation.
- Contains C-terminal His and Avi tags to facilitate purification and immobilization.
- Purity >90% as determined by reducing SDS-PAGE.
- Predicted molecular weight ~55.8 kDa.
- Supplied in PBS, 0.2 μm filtered; avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.