Learn More
Medchemexpress LLC IL-23 Rat HEK293 100ug
SDP
Supplier:  Medchemexpress LLC HYP73195100UG
Encompass_Preferred
IL-23 alpha protein has cytokine activity and binds to the interleukin 23 receptor It regulates cytokine production lymphocyte activation and peptidyl tyrosine phosphorylation IL-23 Protein Rat (HEK293 His) is a recombinant protein dimer complex containing rat-derived IL-23 alpha IL-12 beta Heterodimer protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-81160