Learn More
Medchemexpress LLC IL-23R protein, cynomolgus | 100UG

Supplier: Medchemexpress LLC HYP700889100UG
Recombinant interleukin-23 receptor (IL-23R) extracellular domain from cynomolgus monkey, expressed in HEK293 cells and supplied as a C-terminal His-tagged, lyophilized protein for research applications. Provided in a 100 μg size, the protein shows an apparent molecular weight of 60-80 kDa on SDS-PAGE and has endotoxin <1 EU/μg. Formulated in PBS (pH 7.4) with trehalose and intended for research use only.
- Recombinant extracellular domain expressed in HEK293 cells.
- C-terminal His tag for detection and purification.
- Observed molecular weight 60-80 kDa by SDS-PAGE.
- Endotoxin level <1 EU/μg.
- Lyophilized formulation from PBS, typically with ~8% trehalose.
- Store at -20°C; after reconstitution stable at 4°C for 1 week.
- Recommended to aliquot and store at -20°C or -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.