Learn More
Medchemexpress LLC IL-27 Mouse HEK293 5ug
SDP
Supplier:  Medchemexpress LLC HYP73200Y5UG
Encompass_Preferred
The IL-27 protein together with IL23A forms IL-23 interleukin a key cytokine in innate and adaptive immunity It is associated with infection responses binds to IL12RB1 and IL23R receptor complexes and activates Jak-Stat signaling IL-27 Protein Mouse (228a a HEK293 His solution) is the recombinant mouse-derived IL-27 expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-82273