Learn More
Medchemexpress LLC HY-P7040 10ug Medchemexpress, IL-3, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP704010UG
Encompass_Preferred
Medchemexpress, HY-P7040 10ug IL-3, Human CAS: Recombinant Human Interleukin-3 is a multilineage hematopoietic cytokine with promising effects on platelet and neutrophil counts and special usefulness in patients with secondary hematopoietic failure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3098