Learn More
Medchemexpress LLC IL-3 Human His 100ug

Supplier: Medchemexpress LLC HYP70576100UG
Recombinant human interleukin-3 (IL-3) with an N-terminal His tag, expressed in E. coli and supplied lyophilized. This cytokine supports hematopoietic cell proliferation and is intended for activity, binding, and biochemical assays; reported ED50 in TF-1 proliferation assay is 0.05-0.3 ng/mL. Predicted molecular mass is ~16.6 kDa and purity is >95% by reducing SDS-PAGE.
- Supports TF-1 cell proliferation assays with ED50 0.05-0.3 ng/mL.
- His-tag for easy purification and detection.
- High purity (>95%) verified by reducing SDS-PAGE.
- Lyophilized formulation for improved stability and shipping.
- Store at -20°C for long-term stability; aliquot after reconstitution.
- Includes sequence and molecular mass information to aid assay design.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.