Learn More
Medchemexpress LLC IL-3 Mouse 2ug
SDP
Supplier:  Medchemexpress LLC HYP7062A2UG
Encompass_Preferred
Recombinant mouse interleukin-3 (IL-3), His-tagged, supplied lyophilized in a 2 μg size for research use. IL-3 promotes proliferation, differentiation, and survival of hematopoietic cells and is intended for in vitro laboratory applications. Refer to the manufacturer's datasheet for recommended reconstitution, storage, and handling instructions.
  • Recombinant His-tagged protein expressed in E. coli.
  • Lyophilized format enables long-term storage and convenient reconstitution.
  • Low endotoxin level (<0.2 EU/μg) suitable for sensitive biological assays.
  • Approximate molecular weight ~17 kDa corresponding to the active cytokine domain.
  • Applicable for cell proliferation, differentiation, and functional assays in research.
Catalog No. 50-001-85146