Learn More
Medchemexpress LLC IL-33 Cynomolgus 100ug
SDP
Supplier:  Medchemexpress LLC HYP73883100UG
Encompass_Preferred
IL-33 Protein is a cytokine belonging to the IL-1 superfamily IL-33 induces helper T cells mast cells eosinophils and basophils to produce type 2 cytokines IL-33 acts intracellularly as a nuclear factor and extracellularly as a cytokine IL-33 mediates its biological effects via IL-1 receptor ST 2 activates NF-kappaB and MAP kinases and drives production of T(H)2-associated cytokines from in vitro polarized T(H)2 cells IL-33 Protein Cynomolgus is the recombinant cynomolgus-derived IL-33 protein expressed by E coli with tag free
Catalog No. 50-001-79784