Learn More
Enzo Life Sciences IL-33 (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900201
Encompass_Preferred
The IL-33 (human), EIA kit is a colorimetric immunometric immunoassay kit with results in 3 hours. Alternative name: Interleukin-33, NF-HEV. Sensitivity: 1.7 pg/ml (range 7.8 - 500 pg/ml). Use/Stability: Store all components at +4ºC, except Standard at -20ºC. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-200-6863