Learn More
Medchemexpress LLC IL-36 beta/IL-1F8 H 1mg
SDP
Supplier:  Medchemexpress LLC HYP718621MG
Encompass_Preferred
Recombinant human IL-36 beta (IL-1F8) is a tag-free, lyophilized cytokine produced in E. coli for cell-based assays and biochemical studies. It activates IL-36R to induce NF-κB and MAPK signaling and has verified activity in IL-8 induction assays.
  • Lyophilized, tag-free recombinant protein suitable for reconstitution.
  • Produced in E. coli with a sequence length of 153 amino acids.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~18 kDa under reducing conditions.
  • Verified bioactivity: induces IL-8 in A431 cells (ED50 7.502 ng/mL).
Catalog No. 50-001-83433