Learn More
Medchemexpress LLC IL-36 beta/IL-1F8 H 500ug
SDP
Supplier:  Medchemexpress LLC HYP71862500UG
Encompass_Preferred
Recombinant human IL-36 beta (IL-1F8) is a tag-free, lyophilized cytokine (153 amino acids) produced in E. coli and supplied for in vitro research. It engages the IL-36 receptor to activate NF-κB and MAPK signaling and is intended for receptor-binding, signaling, and cytokine-response assays. The material is provided at high purity and formulated for stability with recommended frozen storage for long-term use.
  • Tag-free recombinant protein expressed in E. coli.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation in PBS for storage stability.
  • Activates IL-36R and downstream NF-κB and MAPK signaling.
  • Suitable for receptor-binding, signaling, and pathway assays.
  • Available in multiple sizes, including a 500 μg option for flexible use.
  • Stable when frozen; aliquot and store at -20°C or -80°C for extended storage.
Catalog No. 50-001-83303