Learn More
Medchemexpress LLC IL-3R alpha/CD123 C 10ug | 10ug
SDP
Supplier:  Medchemexpress LLC HYP7863410UG
Encompass_Preferred
IL-3R /CD123 protein is an important member of the type I cytokine receptor family and mediates cellular responses to a variety of cytokines emphasizing its key role in hematopoiesis and immune regulation IL-3R alpha/CD123 Protein Canine (HEK293 His) is the recombinant canine-derived IL-3R alpha/CD123 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-94116