Learn More
Medchemexpress LLC IL-4 Mouse CHO 2ug
SDP
Supplier:  Medchemexpress LLC HYP70802UG
Encompass_Preferred
Recombinant mouse interleukin-4 (IL-4) produced in CHO cells, provided as a lyophilized preparation for research use. The cytokine promotes proliferation, differentiation, and cell-surface protein modulation of B lymphocytes, and is formulated and stored for stability in laboratory assays and in vitro studies.
  • CHO-derived recombinant protein with species-specific activity.
  • Lyophilized formulation containing trehalose for enhanced stability.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Apparent molecular weight approximately 15-22 kDa due to glycosylation.
  • Recommended storage at -20°C; aliquot and store at -80°C for extended preservation.
Catalog No. 50-001-84966