Learn More
Medchemexpress LLC IL-4 Protein Mouse 100ug
SDP
Supplier:  Medchemexpress LLC HYP73215100UG
Encompass_Preferred
Interleukin 4 (IL-4) Protein is a pleiotropic cytokine secreted primarily by mast cells T-cells eosinophils and basophils that plays a role in regulating antibody production hematopoiesis and inflammation and the development of effector T-cell responses IL-4 participates in STAT6 signaling and regulates the expression of MHC II IgE IgG1 amd CD23 IL-4 Protein Mouse (Q136D Y139D sf9 His) is the recombinant mouse-derived IL-4 protein expressed by Sf9 insect cells with C-His labeled tag and Q136D Y139D mutation
Catalog No. 50-001-82492