Learn More
Enzo Life Sciences IL-7 (mouse), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1380010
Encompass_Preferred
Alternative name: Interleukin-7. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized. Source: Produced in E.coli. Long Term Storage: -20°C.
Catalog No. 50-201-3930