Learn More
ACROBiosystems Recombinant Protein;Human IL-1 Rrp2 / IL-1 R6 Protein;HEK293;1MG;IL2,IL-1 R6,IL-1 Rrp2,IL1RL2,IL-1RL2,IL1Rrp2,IL-1Rrp2,IL1RRP2,IL-36R,IL-36 receptor
Supplier:  ACROBiosystems IL2H52H61MG
Encompass_Preferred
Recombinant Protein;Human IL-1 Rrp2 / IL-1 R6 Protein;1MG;Human IL-1 Rrp2 (IL2-H52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 335 (Accession # Q9HB29-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.8 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7674