Learn More
ACROBiosystems Recombinant Protein;Human IL-2 R beta / CD122 Protein, Fc Tag;HEK293;1MG;ILB,IL2RB,RP5-1170K4.6,CD122,P70-75,IL-2RB,IL2 R Beta,IL-2,IL2
Supplier:  ACROBiosystems ILBH52531MG
Encompass_Preferred
Recombinant Protein;Human IL-2 R beta / CD122 Protein, Fc Tag;1MG;Human IL-2 R beta, Fc Tag (ILB-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Accession # NP_000869.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.7 kDa. The protein migrates as 58-76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-15, Tag Free (Cat. No. IL5-H4217) at 10 μg/mL (100 μL/well) can bind Human IL-2 R beta, Fc Tag (Cat. No. ILB-H5253) with a linear range of 0.156-2.5 μg/mL (QC tested).;Immobilized Human IL-2 R beta, Fc Tag (Cat. No. ILB-H5253) at 2 μg/mL (100 μL/well) can bind Human IL-2, Tag Free (Cat. No. IL2-H4216) with a linear range of 0.156-2.5 μg/mL (Routinely tested).
Catalog No. 50-165-8022