Learn More
Genscript Corporation IL6 55C6 mAb Mouse
Supplier:  Genscript Corporation V064031
Encompass
Interleukin-6 (IL6) is a multi-functional cytokine produced by variety of cells including T-cells B-cells monocytes fibroblasts keratinocytes endothelial cells mesangial cells astrocytes and bone marrow stroma cells It is involved in the induction of B-cell differentiation growth promotion of myeloma cells and proliferation and differentiation of T cells IL6 can regulate immune responses acute phase reactions and hematopoiesis
Catalog No. 50-000-57036